tert-Butyl (2-hydroxyethyl)(methyl)carbamate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N-(tert-Butoxycarbonyl)-N-methylethanolamine, N-Boc-N-methyl-2-aminoethanol, tert-Butyl (2-Hydroxyethyl)(methyl)carbamate, N-(tert-Butoxycarbonyl)-N-methyl-2-aminoethanol, N-Boc-N-methylethanolamine, N-Boc-N-methylglycinol, N-(tert-Butoxycarbonyl)-N-methylglycinol, (2-Hydroxyethyl)(methyl)carbamic acid tert-Butyl ester
Formula:
C₈H₁₇NO₃ Peso molecolare: 175,23 g/mol Punto di ebollizione: 84°C |
MDL Number:
MFCD03425857 CAS Number: 57561-39-4 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...