(E,E)-1,4-Bis[4-[bis(4-methoxyphenyl)amino]styryl]benzene ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
Synonyms:
(E,E)-1,4-Bis[2-[4-[N,N-bis(4-methoxyphenyl)amino]phenyl]vinyl]benzene, (E,E)-4,4'-[1,4-Phenylenebis(ethene-2,1-diyl)]bis[N,N-bis(4-methoxyphenyl)aniline], TOP-HTM-α1
B5672-1G
B5672-25G
B5672-5G
TCIAB5672-1GEA
84
EUR
TCIAB5672-1G
TCIAB5672-25G
TCIAB5672-5G
(E,E)-1,4-Bis[4-[bis(4-methoxyphenyl)amino]styryl]benzene ≥98.0% (by HPLC, total nitrogen)
(E,E)-1,4-Bis[4-[bis(4-methoxyphenyl)amino]styryl]benzene
Formula:
C₅₀H₄₄N₂O₄ Peso molecolare: 736,91 g/mol Punto di fusione: 181°C |
CAS Number:
872466-50-7 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...