Stampa…

(Boc-amino)-PEG4-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Boc-NH-PEG4-CH2CH2COOH, Carboxy-PEG4-(Boc-amine), 15-[(tert-Butoxycarbonyl)amino]-4,7,10,13-tetraoxapentadecanoic acid, 2,2-Dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oic acid, 15-(Boc-amino)-4,7,10,13-tetraoxapentadecanoic acid

B5665-250MG B5665-1G
TCIAB5665-250MGEA 83 EUR
TCIAB5665-250MG TCIAB5665-1G
(Boc-amino)-PEG4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
(Boc-amino)-PEG4-carboxylic acid
Formula: C₁₆H₃₁NO₈
Peso molecolare: 365,42 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD07781254
CAS Number: 756525-91-4

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) (CAD) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR