Stampa…

2-(tert-Butyldimethylsilyl)-N,N-dimethyl-1H-imidazole-1-sulphonamide ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(Dimethylsulphamoyl)-2-(tert-butyldimethylsilyl)-1H-imidazole

B5658-1G B5658-5G
TCIAB5658-1GEA 76 EUR
TCIAB5658-1G TCIAB5658-5G
2-(tert-Butyldimethylsilyl)-N,N-dimethyl-1H-imidazole-1-sulphonamide ≥97.0% (by GC)
2-(tert-Butyldimethylsilyl)-N,N-dimethyl-1H-imidazole-1-sulphonamide
Formula: C₁₁H₂₃N₃O₂SSi
Peso molecolare: 289,47 g/mol
MDL Number: MFCD12963638
CAS Number: 129378-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55a503ba63293d5901f7f30d6ca3444ca27866c7&vid=55a503ba63293d5901f7f30d6ca3444ca27866c7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 62.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR