Stampa…

3-Bromo-N-methylbenzamide ≥98.0% (by GC)

Fornitore: TCI EUROPE

B5644-1G B5644-5G
TCIAB5644-1GEA 57 EUR
TCIAB5644-1G TCIAB5644-5G
3-Bromo-N-methylbenzamide ≥98.0% (by GC)
3-Bromo-N-methylbenzamide
Formula: C₈H₈BrNO
Peso molecolare: 214,0592 g/mol
MDL Number: MFCD01060879
CAS Number: 49834-22-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 91.0 to 95.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR