N,N-Bis(4-bromophenyl)-4-fluoroaniline ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,4'-Dibromo-4''-fluorotriphenylamine, N,N-Bis(4-bromophenyl)-4-fluorobenzenamine
B5637-1G
B5637-5G
TCIAB5637-1GEA
70
EUR
TCIAB5637-1G
TCIAB5637-5G
N,N-Bis(4-bromophenyl)-4-fluoroaniline ≥98.0% (by GC)
N,N-Bis(4-bromophenyl)-4-fluoroaniline
Formula:
C₁₈H₁₂Br₂FN Peso molecolare: 421,11 g/mol |
MDL Number:
MFCD31618116 CAS Number: 1429194-04-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 103.0 to 107.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...