Stampa…

9-[4-(tert-Butyl)phenyl]-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5620-1G B5620-5G
TCIAB5620-1GEA 116 EUR
TCIAB5620-1G TCIAB5620-5G
9-[4-(tert-Butyl)phenyl]-9H-carbazole ≥98.0% (by GC)
9-[4-(tert-Butyl)phenyl]-9H-carbazole
Formula: C₂₂H₂₁N
Peso molecolare: 299,42 g/mol
MDL Number: MFCD31618114
CAS Number: 57103-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3845a8c7ed826d1199620b0333299c18b29196f4&vid=3845a8c7ed826d1199620b0333299c18b29196f4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 189.0 to 193.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR