Stampa…

3-Bromo-9-(2-ethylhexyl)-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5612-1G B5612-5G
TCIAB5612-1GEA 90 EUR
TCIAB5612-1G TCIAB5612-5G
3-Bromo-9-(2-ethylhexyl)-9H-carbazole ≥98.0% (by GC)
3-Bromo-9-(2-ethylhexyl)-9H-carbazole
Formula: C₂₀H₂₄BrN
Peso molecolare: 358,32 g/mol
MDL Number: MFCD18252906
CAS Number: 628336-85-6

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.000 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR