Stampa…

2-Bromo-9-n-octyl-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Bromo-9-N-octyl-9H-carbazole

B5609-1G B5609-5G
TCIAB5609-1GEA 131 EUR
TCIAB5609-1G TCIAB5609-5G
2-Bromo-9-n-octyl-9H-carbazole ≥98.0% (by GC)
2-Bromo-9-n-octyl-9H-carbazole
Formula: C₂₀H₂₄BrN
Peso molecolare: 358,32 g/mol
MDL Number: MFCD31618111
CAS Number: 1356465-23-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=035de448a522b6a8871cf058c5a6517b8720c45c&vid=035de448a522b6a8871cf058c5a6517b8720c45c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR