2-Bromo-9-hexyl-9H-carbazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B5607-1G
B5607-5G
TCIAB5607-1GEA
149
EUR
TCIAB5607-1G
TCIAB5607-5G
2-Bromo-9-hexyl-9H-carbazole ≥98.0% (by GC)
2-Bromo-9-hexyl-9H-carbazole
Formula:
C₁₈H₂₀BrN Peso molecolare: 330,27 g/mol |
MDL Number:
MFCD31618110 CAS Number: 864550-95-8 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 40.0 to 44.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...