Stampa…

2,3-Bis(2,6-di-i-propylphenylimino)butane ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 2,3-Bis(2,6-diisopropylphenylimino)butane, N,N'-Bis(2,6-diisopropylphenyl)butane-2,3-diimine

B5606-200MG B5606-1G
TCIAB5606-200MGEA 125 EUR
TCIAB5606-200MG TCIAB5606-1G
2,3-Bis(2,6-di-i-propylphenylimino)butane ≥98.0% (by HPLC)
2,3-Bis(2,6-di-i-propylphenylimino)butane
Formula: C₂₈H₄₀N₂
Peso molecolare: 404,64 g/mol
MDL Number: MFCD01862435
CAS Number: 74663-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc13f0023780a38d280dae319a94736db27c3638&vid=dc13f0023780a38d280dae319a94736db27c3638&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR