9-([1,1'-Biphenyl]-3-yl)-9H-carbazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B5591-1G
B5591-5G
TCIAB5591-1GEA
42
EUR
TCIAB5591-1G
TCIAB5591-5G
9-([1,1'-Biphenyl]-3-yl)-9H-carbazole ≥98.0% (by GC)
9-([1,1'-Biphenyl]-3-yl)-9H-carbazole
Formula:
C₂₄H₁₇N Peso molecolare: 319,41 g/mol |
MDL Number:
MFCD28167065 CAS Number: 1221237-87-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...