Stampa…

tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate ≥99.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-Boc-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-(Boc-amino)phenylboronic acid Pinacol ester, tert-Butyl [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate, 4-[(tert-Butoxycarbonyl)amino]phenylboronic acid Pinacol ester, 4-Boc-aminophenylboronic acid, pinacol ester, N-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamic acid tert-butyl ester, 4-(Boc-amino)benzeneboronic acid pinacol ester

B5588-1G B5588-5G
TCIAB5588-1GEA 22 EUR
TCIAB5588-1G TCIAB5588-5G
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate ≥99.0% (by HPLC, titration analysis)
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Formula: C₁₇H₂₆BNO₄
Peso molecolare: 319,208 g/mol
MDL Number: MFCD02179439
CAS Number: 330793-01-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f41cc8bdee7eff831405d836c99cd5f8f99d4e0b&vid=f41cc8bdee7eff831405d836c99cd5f8f99d4e0b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR