5-Bromo-3-nitropyridine-2-carbonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
5-Bromo-2-cyano-3-nitropyridine, 5-Bromo-3-nitropicolinonitrile, 5-Bromo-3-nitropyridine-2-carbonitrile
B5584-5G
B5584-25G
TCIAB5584-5GEA
57
EUR
TCIAB5584-5G
TCIAB5584-25G
5-Bromo-3-nitropyridine-2-carbonitrile ≥98.0% (by GC)
5-Bromo-3-nitropyridine-2-carbonitrile
Formula:
C₆H₂BrN₃O₂ Peso molecolare: 228,0030 g/mol |
MDL Number:
MFCD06657551 CAS Number: 573675-25-9 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...