Stampa…

1-Butyl-4-methylpyridiniumtetrafluoroborate ≥98.0% (by HPLC)

Fornitore: TCI
Pericolo

Synonyms: 1-Butyl-4-methylpyridin-1-ium tetrafluoroborate

B5570-25G B5570-5G
TCIAB5570-25GEA 129 EUR
TCIAB5570-25G TCIAB5570-5G
1-Butyl-4-methylpyridiniumtetrafluoroborate ≥98.0% (by HPLC)
1-Butyl-4-methylpyridiniumtetrafluoroborate
Formula: C₁₀H₁₆BF₄N
Punto di fusione: -30°C
MDL Number: MFCD03095460
CAS Number: 343952-33-0
UN: 1760
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR