3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-((tert-Butyldimethylsilyl)oxy)propan-1-ol, 3-[[tert-Butyl(dimethyl)silyl]oxy]propan-1-ol, 3-[(tert-Butyldimethylsilyl)oxy]-1-propanol
B5555-1ML
B5555-5ML
TCIAB5555-1MLEA
37
EUR
TCIAB5555-1ML
TCIAB5555-5ML
3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol ≥96.0% (by GC)
3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol
Formula:
C₉H₂₂O₂Si Peso molecolare: 190,36 g/mol Flash Pt: 93°C |
MDL Number:
MFCD00187968 CAS Number: 73842-99-6 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...