9-(6-Bromohexyl)-9H-carbazole ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B5548-200MG
B5548-1G
TCIAB5548-200MGEA
78
EUR
TCIAB5548-200MG
TCIAB5548-1G
9-(6-Bromohexyl)-9H-carbazole ≥97.0% (by GC)
9-(6-Bromohexyl)-9H-carbazole
Formula:
C₁₈H₂₀BrN Peso molecolare: 330,27 g/mol |
MDL Number:
MFCD30562129 CAS Number: 94847-10-6 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...