Stampa…

9-(4-Bromobutyl)-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI
Attenzione

B5547-200MG B5547-1G
TCIAB5547-200MGEA 50 EUR
TCIAB5547-200MG TCIAB5547-1G
9-(4-Bromobutyl)-9H-carbazole ≥98.0% (by GC)
9-(4-Bromobutyl)-9H-carbazole
Formula: C₁₆H₁₆BrN
MDL Number: MFCD11546053
CAS Number: 10420-20-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR