Bis(4-bromophenyl)phenylphosphine oxide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B5532-200MG
B5532-1G
TCIAB5532-200MGEA
39
EUR
TCIAB5532-200MG
TCIAB5532-1G
Bis(4-bromophenyl)phenylphosphine oxide ≥98.0% (by GC)
Bis(4-bromophenyl)phenylphosphine oxide
Formula:
C₁₈H₁₃Br₂OP Peso molecolare: 436,08 g/mol Punto di ebollizione: 280°C |
CAS Number:
93869-52-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...