Bis(3-bromophenyl)phenylphosphine oxide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B5531-1G
B5531-200MG
TCIAB5531-1GEA
151
EUR
TCIAB5531-1G
TCIAB5531-200MG
Bis(3-bromophenyl)phenylphosphine oxide ≥98.0% (by GC)
Bis(3-bromophenyl)phenylphosphine oxide
Formula:
C₁₈H₁₃Br₂OP Peso molecolare: 436,08 g/mol |
CAS Number:
1163698-32-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 108.0 to 112.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...