Stampa…

(3-Bromophenyl)diphenylphosphine oxide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5530-200MG B5530-1G
TCIAB5530-200MGEA 38 EUR
TCIAB5530-200MG TCIAB5530-1G
(3-Bromophenyl)diphenylphosphine oxide ≥98.0% (by GC)
(3-Bromophenyl)diphenylphosphine oxide
Formula: C₁₈H₁₄BrOP
Peso molecolare: 357,19 g/mol
MDL Number: MFCD30533951
CAS Number: 10212-04-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1f3b3f0af2c09519a9195aadaa3a6e7b9473f87&vid=a1f3b3f0af2c09519a9195aadaa3a6e7b9473f87&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR