Stampa…

1-Butyl-3-methylimidazolium methyl sulphate ≥98.0% (by HPLC)

Fornitore: TCI
Pericolo

Synonyms: [BMIM] [MeSO4], 1-n-Butyl-3-methylimidazolium methyl sulphate, ECOENG 411, BMIMMeSO4

B5527-5G B5527-25G
TCIAB5527-5GEA 70 EUR
TCIAB5527-5G TCIAB5527-25G
1-Butyl-3-methylimidazolium methyl sulphate ≥98.0% (by HPLC)
1-Butyl-3-methylimidazolium methyl sulphate
Formula: C₉H₁₈N₂O₄S
Peso molecolare: 250.32 g/mol
Punto di fusione: –4°C
Densità: 1.21
MDL Number: MFCD03095437
CAS Number: 401788-98-5
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ec86c0fdf953dff3d72ad43639454fbf76b46d7&vid=1ec86c0fdf953dff3d72ad43639454fbf76b46d7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR