Stampa…

2-Bromo-4-chlorobenzaldehyde ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5524-5G B5524-25G
TCIAB5524-5GEA 42 EUR
TCIAB5524-5G TCIAB5524-25G
2-Bromo-4-chlorobenzaldehyde ≥97.0% (by GC)
2-Bromo-4-chlorobenzaldehyde
Formula: C₇H₄BrClO
MDL Number: MFCD09056792
CAS Number: 84459-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c9809f02811ba0dfabd807d3d44588f8bdb5f98&vid=7c9809f02811ba0dfabd807d3d44588f8bdb5f98&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR