Stampa…

BMS-599626 ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: (S)-Morpholin-3-ylmethyl [4-[[1-(3-Fluorobenzyl)-1H-indazol-5-yl]amino]-5-methylpyrrolo[2,1-f][1,2,4]triazin-6-yl]carbamate, AC 480

B5520-10MG B5520-50MG
TCIAB5520-10MGEA 209 EUR
TCIAB5520-10MG TCIAB5520-50MG
BMS-599626 ≥98.0% (by HPLC)
BMS-599626
Formula: C₂₇H₂₇FN₈O₃
Peso molecolare: 530,6 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD10565678
CAS Number: 714971-09-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2b324f6da0356c0e76bd047fe28ad44dd7cc263&vid=e2b324f6da0356c0e76bd047fe28ad44dd7cc263&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 20.00 to 22.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR