Stampa…

tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate ≥97.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: tert-Butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate, 1-Oxa-6-azaspiro[2.5]octane, N-BOC protected

B5501-1G B5501-5G
TCIAB5501-1GEA 49 EUR
TCIAB5501-1G TCIAB5501-5G
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate ≥97.0% (by GC)
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate
Formula: C₁₁H₁₉NO₃
Peso molecolare: 213,28 g/mol
MDL Number: MFCD07779385
CAS Number: 147804-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0afa8382852b19ee12407a8fb0246b110c37c0e5&vid=0afa8382852b19ee12407a8fb0246b110c37c0e5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 55.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR