Stampa…

4,8-Bis[(2-ethylhexyl)oxy]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene ≥97.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: [4,8-Bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b']dithiophene-2,6-diyl]bis(trimethylstannane), (4,8-Bis((2-ethylhexyl)oxy)benzo[1,2-b:4,5-b']dithiophene-2,6-diyl)bis(trimethylstannane)

B5490-500MG B5490-5G
TCIAB5490-500MGEA 97 EUR
TCIAB5490-500MG TCIAB5490-5G
4,8-Bis[(2-ethylhexyl)oxy]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene ≥97.0% (by HPLC)
4,8-Bis[(2-ethylhexyl)oxy]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene
Formula: C₃₂H₅₄O₂S₂Sn₂
Peso molecolare: 772,32 g/mol
MDL Number: MFCD20275089
CAS Number: 1160823-78-8
UN: 3146
ADR: 6.1,I

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) min. 97.0 area%
Elemental analysis (Sulfuride) 7.80 to 8.80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR