Stampa…

1-Butyl-3-methylimidazolium tricyanomethanide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

B5487-5G B5487-25G
TCIAB5487-5GEA 51 EUR
TCIAB5487-5G TCIAB5487-25G
1-Butyl-3-methylimidazolium tricyanomethanide ≥98.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazolium tricyanomethanide
Formula: C₁₂H₁₅N₅
Peso molecolare: 229,29 g/mol
MDL Number: MFCD09038877
CAS Number: 878027-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c30d8a74ac480894af052e5b0e5f4870157cadb2&vid=c30d8a74ac480894af052e5b0e5f4870157cadb2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR