AlkylFluor(TM) ≥97.0% (by total nitrogen basis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₂₇H₃₆BF₅N₂ |
CAS Number:
2043361-32-4 UN: 1759 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...