Stampa…

Bicyclo[1.1.1]pentan-1-amine hydrochloride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B5479-200MG B5479-1G
TCIAB5479-200MGEA 78 EUR
TCIAB5479-200MG TCIAB5479-1G
Bicyclo[1.1.1]pentan-1-amine hydrochloride ≥98.0% (by GC, titration analysis)
Bicyclo[1.1.1]pentan-1-amine hydrochloride
Formula: C₅H₉N·HCl
Peso molecolare: 119,59 g/mol
MDL Number: MFCD28166320
CAS Number: 22287-35-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79ce607854801725e63dc9c82789f2edf0e7685f&vid=79ce607854801725e63dc9c82789f2edf0e7685f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR