Stampa…

2-Bromo-5-(2-ethylhexyl)thiophene ≥98.0% (by GC)

Fornitore: TCI EUROPE

B5475-1G B5475-5G
TCIAB5475-1GEA 120 EUR
TCIAB5475-1G TCIAB5475-5G
2-Bromo-5-(2-ethylhexyl)thiophene ≥98.0% (by GC)
2-Bromo-5-(2-ethylhexyl)thiophene
Formula: C₁₂H₁₉BrS
Peso molecolare: 275,25 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD28101692
CAS Number: 925899-21-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR