Stampa…

3-(2-Bromophenyl)-9-phenyl-9H-carbazole ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5465-1G B5465-5G
TCIAB5465-1GEA 165 EUR
TCIAB5465-1G TCIAB5465-5G
3-(2-Bromophenyl)-9-phenyl-9H-carbazole ≥97.0% (by GC)
3-(2-Bromophenyl)-9-phenyl-9H-carbazole
Formula: C₂₄H₁₆BrN
Peso molecolare: 398,30 g/mol
MDL Number: MFCD29071836
CAS Number: 1190100-35-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a2520ae058005c039bb0d0257b54303d68d064e&vid=1a2520ae058005c039bb0d0257b54303d68d064e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 122.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR