Stampa…

2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]malononitrile ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]propanedinitrile

B5458-200MG B5458-1G
TCIAB5458-200MGEA 75 EUR
TCIAB5458-200MG TCIAB5458-1G
2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]malononitrile ≥95.0% (by GC)
2-[(5'-Bromo-[2,2'-bithiophen]-5-yl)methylene]malononitrile
Formula: C₁₂H₅BrN₂S₂
Peso molecolare: 321,21 g/mol
MDL Number: MFCD06214120
CAS Number: 704890-84-6

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 95.0 %
Melting point 200.0 to 204.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR