Stampa…

5'-Bromo-[2,2'-bithiophene]-5-carbonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5457-200MG B5457-1G
TCIAB5457-200MGEA 79 EUR
TCIAB5457-200MG TCIAB5457-1G
5'-Bromo-[2,2'-bithiophene]-5-carbonitrile ≥98.0% (by GC)
5'-Bromo-[2,2'-bithiophene]-5-carbonitrile
Formula: C₉H₄BrNS₂
Peso molecolare: 270,16 g/mol
CAS Number: 176787-96-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aa72c5e7eeab656e583c1346167eb3be7880ade&vid=5aa72c5e7eeab656e583c1346167eb3be7880ade&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 161.0 to 165.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR