Stampa…

Bis[2-[(oxo)diphenylphosphino]phenyl] ether ≥99.0% (by HPLC), purified by sublimation

Fornitore: TCI EUROPE

Synonyms: (Oxybis(2,1-phenylene))bis(diphenylphosphine oxide), DPEPO

B5448-200MG B5448-1G
TCIAB5448-200MGEA 60 EUR
TCIAB5448-200MG TCIAB5448-1G
Bis[2-[(oxo)diphenylphosphino]phenyl] ether ≥99.0% (by HPLC), purified by sublimation
Bis[2-[(oxo)diphenylphosphino]phenyl] ether
Formula: C₃₆H₂₈O₃P₂
Peso molecolare: 570,56 g/mol
Punto di fusione: 280°C
MDL Number: MFCD28968111
CAS Number: 808142-23-6

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) min. 99.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR