Stampa…

1-Bromo-3,5-diethynylbenzene ≥98.0% (by GC)

Fornitore: TCI

B5444-1G B5444-200MG
TCIAB5444-1GEA 227 EUR
TCIAB5444-1G TCIAB5444-200MG
1-Bromo-3,5-diethynylbenzene ≥98.0% (by GC)
1-Bromo-3,5-diethynylbenzene
Formula: C₁₀H₅Br
MDL Number: MFCD27665569
CAS Number: 144001-08-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR