Stampa…

2-(2-Bromophenyl)-9-phenyl-9H-carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5437-1G B5437-200MG
TCIAB5437-1GEA 297 EUR
TCIAB5437-1G TCIAB5437-200MG
2-(2-Bromophenyl)-9-phenyl-9H-carbazole ≥98.0% (by GC)
2-(2-Bromophenyl)-9-phenyl-9H-carbazole
Formula: C₂₄H₁₆BrN
Peso molecolare: 398,30 g/mol
MDL Number: MFCD27644694
CAS Number: 1616607-88-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR