Stampa…

BTT 3033 ≥96.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 1-(4-Fluorophenyl)-N-methyl-N-(4-(3-phenylureido)phenyl)-1H-pyrazole-4-sulphonamide, BTT-3033

B5415-10MG B5415-50MG
TCIAB5415-10MGEA 232 EUR
TCIAB5415-10MG TCIAB5415-50MG
BTT 3033 ≥96.0% (by HPLC)
BTT 3033
Formula: C₂₃H₂₀FN₅O₃S
Peso molecolare: 465,50 g/mol
MDL Number: MFCD28166480
CAS Number: 1259028-99-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f824f530cbc7e9d0ced54b977d48c83922c1f24e&vid=f824f530cbc7e9d0ced54b977d48c83922c1f24e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 96.0 area%
Elemental analysis (Nitrogen) 14.00 to 16.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR