2-Bromo-5-decylthiophene ≥97.0% (by GC)
Fornitore: TCI EUROPE
B5406-1G
B5406-5G
TCIAB5406-1GEA
234
EUR
TCIAB5406-1G
TCIAB5406-5G
2-Bromo-5-decylthiophene ≥97.0% (by GC)
2-Bromo-5-decylthiophene
Formula:
C₁₄H₂₃BrS Peso molecolare: 303,30 g/mol Punto di ebollizione: 135°C |
CAS Number:
514188-72-8 |
Risultati del test di specificazione
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...