Stampa…

2-Bromoindolo[3,2,1-jk]carbazole ≥98.0% (by GC)

Fornitore: TCI

B5397-200MG B5397-1G
TCIAB5397-200MGEA 56 EUR
TCIAB5397-200MG TCIAB5397-1G
2-Bromoindolo[3,2,1-jk]carbazole ≥98.0% (by GC)
2-Bromoindolo[3,2,1-jk]carbazole
Formula: C₁₈H₁₀BrN
MDL Number: MFCD30187281
CAS Number: 1174032-81-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 216.0 to 220.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR