1,1-Bis(hydroxymethyl)cyclopropane ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1,1-Bis(Hydroxymethyl)cyclopropane
B5365-5G
B5365-25G
TCIAB5365-5GEA
21
EUR
TCIAB5365-5G
TCIAB5365-25G
1,1-Bis(hydroxymethyl)cyclopropane ≥96.0% (by GC)
1,1-Bis(hydroxymethyl)cyclopropane
Formula:
C₅H₁₀O₂ Peso molecolare: 102,133 g/mol Punto di ebollizione: 98°C |
MDL Number:
MFCD00464207 CAS Number: 39590-81-3 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...