1-Bromo-2-chloro-4-nitrobenzene ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₃BrClNO₂ |
MDL Number:
MFCD00051514 CAS Number: 29682-39-1 EINECS: 249-773-8 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...