Stampa…

3-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥98.0% (by GC)

Fornitore: TCI EUROPE

B5333-5G B5333-25G
TCIAB5333-5GEA 41 EUR
TCIAB5333-5G TCIAB5333-25G
3-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥98.0% (by GC)
3-Bromo-α,α,α-trifluoro-5-nitrotoluene
Formula: C₇H₃BrF₃NO₂
Peso molecolare: 270,01 g/mol
MDL Number: MFCD03788719
CAS Number: 630125-49-4

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR