Stampa…

3-Bromo-4'-iodo-1,1'-biphenyl ≥95.0% (by GC)

Fornitore: TCI EUROPE

B5321-200MG B5321-1G
TCIAB5321-200MGEA 49 EUR
TCIAB5321-200MG TCIAB5321-1G
3-Bromo-4'-iodo-1,1'-biphenyl ≥95.0% (by GC)
3-Bromo-4'-iodo-1,1'-biphenyl
Formula: C₁₂H₈BrI
Peso molecolare: 359,00 g/mol
MDL Number: MFCD28952954
CAS Number: 187275-73-6
UN: 3152
ADR: 9,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ca9135fc34ef4bf4545bdadc9fcec9bc2f07d9d&vid=4ca9135fc34ef4bf4545bdadc9fcec9bc2f07d9d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 95.0 %
Melting point 45.0 to 49.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR