trans,trans-4'-Butylbicyclohexyl-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₇H₃₀O₂ |
MDL Number:
MFCD06797517 CAS Number: 89111-63-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Mesomorphic range | 211 to 243 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...