Stampa…

(S)-3-(Benzoylthio)-2-methylpropionic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B5315-1G B5315-5G
TCIAB5315-1GEA 68 EUR
TCIAB5315-1G TCIAB5315-5G
(S)-3-(Benzoylthio)-2-methylpropionic acid ≥98.0% (by GC, titration analysis)
(S)-3-(Benzoylthio)-2-methylpropionic acid
Formula: C₁₁H₁₂O₃S
Peso molecolare: 224,27 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00038376
CAS Number: 72679-02-8
EINECS: 276-764-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f14f446fb0ce873315ace17c5e87159f86e0c6f2&vid=f14f446fb0ce873315ace17c5e87159f86e0c6f2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C
Specific rotation [a]20/D -41.0 to -45.0 deg (C=1,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR