Stampa…

9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-(4-Bromophenyl)-3,6-di-tert-butyl-9H-carbazole

B5313-1G B5313-5G
TCIAB5313-1GEA 97 EUR
TCIAB5313-1G TCIAB5313-5G
9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole ≥98.0% (by GC)
9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole
Formula: C₂₆H₂₈BrN
Peso molecolare: 434,42 g/mol
MDL Number: MFCD11617971
CAS Number: 601454-33-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3240eff02ecc79b41fa7be58a41b0b17c7bbb1ae&vid=3240eff02ecc79b41fa7be58a41b0b17c7bbb1ae&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR