Stampa…

1-Boc-3-carbamoylpiperidine ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: tert-Butyl 3-Carbamoyl-1-piperidinecarboxylate, 1-Boc-3-piperidinecarboxamide, 3-Carbamoyl-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-3-Carbamoylpiperidine, 1-(tert-Butoxycarbonyl)-3-piperidinecarboxamide

B5292-1G B5292-5G
TCIAB5292-1GEA 37 EUR
TCIAB5292-1G TCIAB5292-5G
1-Boc-3-carbamoylpiperidine ≥98.0% (by GC)
1-Boc-3-carbamoylpiperidine
Formula: C₁₁H₂₀N₂O₃
Peso molecolare: 228,2881 g/mol
MDL Number: MFCD06658365
CAS Number: 91419-49-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 174.0 to 178.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR