6-Bromo-2(3H)-benzoxazolone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Bromo-2-benzoxazolinone, 6-Bromo-2-benzoxazolone, 6-Bromo-2-hydroxybenzoxazole, 6-Bromobenzo[d]oxazol-2(3H)-one
B5290-1G
B5290-5G
TCIAB5290-1GEA
52
EUR
TCIAB5290-1G
TCIAB5290-5G
6-Bromo-2(3H)-benzoxazolone ≥98.0% (by GC)
6-Bromo-2(3H)-benzoxazolone
Formula:
C₇H₄BrNO₂ Peso molecolare: 214,018 g/mol |
MDL Number:
MFCD00694727 CAS Number: 19932-85-5 |
Risultati del test di specificazione
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 193.0 to 197.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...