4-Bromo-3-methylbenzyl alcohol ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
(4-Bromo-3-methylphenyl)methanol
B5285-1G
B5285-5G
TCIAB5285-1GEA
74
EUR
TCIAB5285-1G
TCIAB5285-5G
4-Bromo-3-methylbenzyl alcohol ≥98.0% (by GC)
4-Bromo-3-methylbenzyl alcohol
Formula:
C₈H₉BrO Peso molecolare: 201,06 g/mol Punto di ebollizione: 90°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD09264005 CAS Number: 149104-89-2 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...