Stampa…

3-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B5273-1G B5273-5G
TCIAB5273-1GEA 49 EUR
TCIAB5273-1G TCIAB5273-5G
3-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC)
3-Bromo-2-hydroxybenzonitrile
Formula: C₇H₄BrNO
Peso molecolare: 198,02 g/mol
MDL Number: MFCD11042854
CAS Number: 13073-28-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR